MultiScreen Filter Plates with Immobilon™-P PVDF Membrane, 96-Well
Leverancier: Merck
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
MAIPN0B50 MAIPN4550 MAIPS4510 MSIPS4510 MSIPN4550 MSIPS4W10 MSIPN4510 MSIPN4W50 MSIPN4B50 S2EM004M99 MSIPN4B10
MILFMAIPN0B50EA 1720 EUR
MILFMAIPN0B50 MILFMAIPN4550 736-0188 MILFMSIPS4510 MILFMSIPN4550 MILFMSIPS4W10 MILFMSIPN4510 MILFMSIPN4W50 MILFMSIPN4B50 MILFS2EM004M99 MILFMSIPN4B10
MultiScreen Filter Plates with Immobilon™-P PVDF Membrane, 96-Well
Microplaten Filtreerplaten
MultiScreen HTS 96-well filtration plates are used to filter samples and perform entire procedures, from cell growth to scintillation counting within the same plate.
  • Hydrophobic Immobilon®-P PVDF membrane
The plates come in many membrane types, pore sizes, and plate materials. The automation-compatible design allows for easy manipulation with a variety of robotics systems and makes barcoding possible on all four sides. Filtration is accomplished either by vacuum using the MultiScreen HTS Vacuum Manifold, or by centrifugation.
MultiScreenHTS platesMore Product Information