Cellulose Fiber Membrane Filters
Leverancier: Merck
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
AP1001300 AP1003700 AP1004700 AP30034P0
MILFAP1001300EA 46.7 EUR
MILFAP1001300 MILFAP1003700 MILFAP1004700 MILFAP30034P0
Cellulose Fiber Membrane Filters
Filters Membraanfilters
Cellulose support pads are used to reinforce filters in monitors for contamination analysis. When saturated with growth medium, they can also be used for culturing microorganisms on membranes.
  • Filter code: AP10
  • Filter type: Depth filter
  • Hydrophilic
Applications include: Air monitoring, environmental monitoring, aerosol contamination monitoring, protecting membrane filters during high pressure or fast flow conditions.
Verpakking: Comes in a pack of 100.