Millicell® High Yield Multilayer Culture Flasks
Leverancier: Merck
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
PFHYS1008 PFHYS0616
MILFPFHYS1008EA 751 EUR
MILFPFHYS1008 MILFPFHYS0616
Millicell® High Yield Multilayer Culture Flasks
Kolven Celcultuurflessen
When growing large numbers of cells, you need a high throughput flask to save space and time. Each Millicell HY (high yield) flask provides a consistent, quality growth environment across all layers, with the same volume of media in each layer. Recovering cells is as easy as growing them.
  • Consistent, high cell yields and viability
  • Same media requirements (volume per unit area) as traditional T-flasks
  • Uniform culture conditions without media leaking or spillover between layers
With easy access to pipetting or pouring, Millicell HY flasks deliver reproducible, high yields of healthy cells. Plus, they’ve been tested for use with stem cells.