EZ-Fit™ Filtration Unit
Leverancier: Merck
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
EFHAB100B EFHAB100I EFHAB250B EFHAB250I EFHAW100B
MILFEFHAB100BEA 0 EUR
MILFEFHAB100B MILFEFHAB100I MILFEFHAB250B MILFEFHAB250I MILFEFHAW100B
EZ-Fit™ Filtration Unit
Filters Flesfilters
The EZ-Fit™ Filtration unit is a disposable filtration device for bioburden testing of liquid samples including water, in-process samples, and final products.
  • Drain support and optimized funnel-to-base assembly ensure optimal microbial recovery
  • Reduced risk of contaminating the membrane with forceps due to a protective rim
  • Visibility of the end of the filtration from any angle thanks to transparent funnel material
  • All around level volume indicators
  • Funnel handling with a single hand
  • Stackability, saving space in the work area
The EZ-Fit™ Filtration Units have been designed to ensure optimal microbial recovery. The sterilized, disposable filtration devices for bioburden testing streamline your workflow, save time and give you highly reliable microbial enumeration results.The blue and pink drain support and the funnel-to-base assembly have been optimized so that the membrane remains perfectly flat after the sample filtration. When transferred to the culture medium, the whole filtration surface of the membrane is in perfect contact with the culture media. The shape of the funnel and the hydrophobic material minimize sample residue ensuring that the complete sample volume reaches the membrane surface. The innovative funnel assembly design prevents any leaks or sample by-pass of the membrane, meaning that the whole sample volume is filtered. The EZ-Fit™ Filtration Units conform to international standards (EP/USP) and comply with water testing regulations.
Key applications: General microbiological analysis, Clinical, Cosmetics, Environmental, Food and beverage, Industrial QC and Pharmaceutical
Funnel material: Styrene butadiene copolymer (SBC)
Filter diameter: 47 mm
Filter surface: Gridded