MF-Millipore™ Membrane Filters, White
Leverancier: Merck
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
HAWP00010 HAWP304F0 HAWG047500
MILFHAWP00010EA 462 EUR
MILFHAWP00010 MILFHAWP304F0 MILFHAWG047500
MF-Millipore™ Membrane Filters, White
Filters Membraanfilters
Biologically inert mixtures of cellulose acetate and cellulose nitrate have made MF-Millipore™ membrane filters one of the most widely used membranes in analytical and research applications.
  • Versatile filter for biological and environmental monitoring applications
  • Available in a range of pore sizes, colored black or white, with or without a gridded surface
  • Compatible with ethylene oxide, gamma irradiation, and autoclave sterilization methods
MF-Millipore™ filters without Triton® surfactant contain minimum amounts of wetting agent and have a lower water extractable content than standard MF-Millipore™ filters.