Empty Monitor Cassettes
Leverancier: Merck
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
M000025A0 M00003700 M000037A0
MILFM000025A0EA 436 EUR
MILFM000025A0 MILFM00003700 MILFM000037A0
Empty Monitor Cassettes
Filters Cassettefilters
  • Material: Polystyrene
  • Non sterile
  • Female Luer inlet slip; Female Luer outlet slip