Microfil® Funnel and Filters
Leverancier: Merck
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
MIHAWG250 MIGSWG250
MILFMIHAWG250EA 384 EUR
MILFMIHAWG250 MILFMIGSWG250
Microfil® Funnel and Filters
Trechters Filtertrechters Filtertrechters
Microfil® funnels and filters assuring leak-free operation and uniform microorganism recoveries as well as an easy transfer of the membrane with forceps and prevents accidentally touching the filtration area.
  • Filter color: White
  • Filter surface: Gridded
  • Chemistry: Mixed Cellulose Esters (MCE)
  • Sterilised
Application: Specific test regulations (clinical, cosmetics, environmental, food and beverage, industrial QC).