Millex® Syringe Filters, PVDF Membrane
Leverancier: Merck
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
SLGVS25LS SLGVS25XS SLGVS25PS SLGVS25US
MILFSLGVS25LSEA 384 EUR
MILFSLGVS25LS MILFSLGVS25XS MILFSLGVS25PS MILFSLGVS25US
Millex® Syringe Filters, PVDF Membrane
Filters Spuitfilters
Syringe filters with hydrophobic PVDF membrane. Used in vacuum line protection, transducer protection and gas filtration. Available as either 25 mm or pump-driven Millex®-FG₅₀ 50 mm diameter filters.
Filters are either non sterile or sterilised with ethylene oxide.