Millex® Syringe Filters, HPF, Nylon Membrane with Glass Fibre Pre-filter
Leverancier: Merck
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
SLHNMZ5NZ SLHNM25NS SLHNM25NK SLGNM25NK SLGNM25NS
514-0436EA 1030 EUR
514-0436 514-0435 MILFSLHNM25NK MILFSLGNM25NK MILFSLGNM25NS
Millex® Syringe Filters, HPF, Nylon Membrane with Glass Fibre Pre-filter
Filters Spuitfilters
These filters include multiple media: a graduated prefilter and a membrane filter for high throughput filtration of viscous or particle-laden solutions.
  • High-density polyethylene housing compatible with the most commonly used solvents
  • Multi-layer filter can improve throughput of high particulate/viscous samples
Applications include analytical sample preparation, wine analysis and general particulate removal.