Millex® Syringe Filter Units, Hydrophobic PTFE Membrane
Leverancier: Merck
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
SLFH025NS SLFH025NB SLFG025NB SLFG025NK SLFH025NK SLLS025NS SLFG025NS SLFHR04NL SLFGR04NL SLFGX13NK SLFGX13NL SLFGX13TL SLFHX13NK SLFHX13NL SLFHX13TL
514-0417EA 316 EUR
514-0417 516-6658 516-6659 MILFSLFG025NK MILFSLFH025NK MILFSLLS025NS 514-0411 514-0418 514-0414 MILFSLFGX13NK MILFSLFGX13NL MILFSLFGX13TL MILFSLFHX13NK MILFSLFHX13NL MILFSLFHX13TL
Millex® Syringe Filter Units, Hydrophobic PTFE Membrane
Filters Spuitfilters
HPLC certified filters for the filtration of organic and aqueous solutions in HPLC.
  • Low protein binding and high chemical resistance
Tested and certified for UV-absorbing substances at wavelengths of 210 and 254 nm with water and acetonitrile; ensuring consistent quality between batches.
Inlet/outlet: Luer lock female/Luer male.