Millex® Syringe Filter Units, Nylon Membrane
Leverancier: Merck
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
SLGN033NB SLGN033NS SLHN033NK SLGN033NK SLHN033NS SLHN033NB SLHNX13NL SLGNX13NK SLGNX13NL SLGNX13TL SLHNX13NK SLHNX13TL
MILFSLGN033NBEA 882 EUR
MILFSLGN033NB MILFSLGN033NS MILFSLHN033NK MILFSLGN033NK MILFSLHN033NS MILFSLHN033NB MILFSLHNX13NL MILFSLGNX13NK MILFSLGNX13NL MILFSLGNX13TL MILFSLHNX13NK MILFSLHNX13TL
Millex® Syringe Filter Units, Nylon Membrane
Filters Spuitfilters
HPLC certified filters for the filtration of organic and aqueous solutions in HPLC.
  • Low protein binding and high chemical resistance
Tested and certified for UV-absorbing substances at wavelengths of 210 and 254 nm with water and acetonitrile; ensuring consistent quality between batches.
Inlet/outlet: Luer lock female/Luer male.