Millex® Syringe Filter Units, PES Membrane
Leverancier: Merck
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
SLGP033NK SLGP033NB SLGP033NS SLHP033NB SLHP033NK SLHP033NS SLGPR33RS
MILFSLGP033NKEA 2200 EUR
MILFSLGP033NK MILFSLGP033NB MILFSLGP033NS MILFSLHP033NB MILFSLHP033NK MILFSLHP033NS MILFSLGPR33RS
Millex® Syringe Filter Units, PES Membrane
Filters Spuitfilters
HPLC certified filters for the filtration of organic and aqueous solutions in HPLC.
  • Low protein binding and high chemical resistance
Tested and certified for UV-absorbing substances at wavelengths of 210 and 254 nm with water and acetonitrile; ensuring consistent quality between batches.
Inlet/outlet: Luer lock female/Luer male.