Millex® Syringe Filter Units, PVDF Membrane
Leverancier: Merck
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
SLHVR04NL SLHVR04NK SLGVR04NL SLGVR04NK SLHV033NK SLHV033NS SLHV033NB SLGV033NK SLGV033NB SLGV033NS SLHVX13NL SLGVX13NK SLGVX13NL SLGVX13TL SLHVX13NK SLHVX13TL
514-0446EA 241 EUR
514-0446 MILFSLHVR04NK 514-0430 514-0429 MILFSLHV033NK MILFSLHV033NS MILFSLHV033NB MILFSLGV033NK MILFSLGV033NB MILFSLGV033NS MILFSLHVX13NL MILFSLGVX13NK MILFSLGVX13NL MILFSLGVX13TL MILFSLHVX13NK MILFSLHVX13TL
Millex® Syringe Filter Units, PVDF Membrane
Filters Spuitfilters
HPLC certified filters for the filtration of organic and aqueous solutions in HPLC.
  • Low protein binding and high chemical resistance
Tested and certified for UV-absorbing substances at wavelengths of 210 and 254 nm with water and acetonitrile; ensuring consistent quality between batches.
Inlet/outlet: Luer lock female/Luer male.