Biomax® PB Ultrafiltration Membrane Disc Filters
Leverancier: Merck
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
PBCC02510 PBCC04310 PBCC04710 PBCC06210 PBCC07610 PBCC09005 PBCC15005 PBGC02510 PBGC04310 PBGC04710 PBGC06210 PBGC07610 PBGC09005 PBGC15005 PBTK02510 PBTK04310 PBTK04710 PBTK06210 PBTK07610 PBTK15005 PBQK02510 PBQK04310 PBQK04710 PBQK06210 PBQK07610 PBQK09005 PBQK15005 PBHK02510 PBHK04310 PBHK04710 PBHK06210 PBHK07610 PBHK15005 PBMK02510 PBMK04310 PBMK04710 PBMK06210 PBMK07610 PBMK09005 PBMK15005 PBVK02510 PBVK04310 PBVK04710 PBVK06210 PBVK07610 PBVK15005
MILFPBCC02510EA 214 EUR
MILFPBCC02510 MILFPBCC04310 MILFPBCC04710 MILFPBCC06210 MILFPBCC07610 MILFPBCC09005 MILFPBCC15005 MILFPBGC02510 MILFPBGC04310 MILFPBGC04710 MILFPBGC06210 MILFPBGC07610 MILFPBGC09005 MILFPBGC15005 MILFPBTK02510 MILFPBTK04310 MILFPBTK04710 MILFPBTK06210 MILFPBTK07610 MILFPBTK15005 MILFPBQK02510 MILFPBQK04310 MILFPBQK04710 MILFPBQK06210 MILFPBQK07610 MILFPBQK09005 MILFPBQK15005 MILFPBHK02510 MILFPBHK04310 MILFPBHK04710 MILFPBHK06210 MILFPBHK07610 MILFPBHK15005 MILFPBMK02510 MILFPBMK04310 MILFPBMK04710 MILFPBMK06210 MILFPBMK07610 MILFPBMK09005 MILFPBMK15005 MILFPBVK02510 MILFPBVK04310 MILFPBVK04710 MILFPBVK06210 MILFPBVK07610 MILFPBVK15005
Biomax® PB Ultrafiltration Membrane Disc Filters
Filters Membraanfilters
Ultrafiltration cut disc membranes are available in Ultracel® regenerated cellulose and Biomax® polyethersulvone (PES).
Ultracel regenerated cellulose membranes are recommended for concentrating or desalting dilute solutions. The hydrophilic and tight microstructure assures the highest possible retention with the lowest possible adsorption of protein, DNA or other macromolecules.
Biomax PES membranes are recommended for concentrating or desalting higher volumes of more concentrated samples such as serum, plasma, or conditioned tissue culture media.