Starter-Kit, SL 1011
Leverancier: EBRO
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
1340-5831 1340-6193 1250-1011
EBRO1340-5831EA 107 EUR
EBRO1340-5831 EBRO1340-6193 EBRO1250-1011
Starter-Kit, SL 1011
Dataloggers
The SL1011 set by ebro, featuring the EBI 12-T101 data logger, PHX800 pH tester, and TDS3 conductivity meter, is designed for validating and monitoring bedpan washers in healthcare settings, ensuring hygiene standards.
  • Comprehensive validation set for bedpan washers
  • Includes EBI 12-T101 temperature data logger, PHX800 pH tester, and TDS3 conductivity meter
  • High-precision monitoring for temperature, pH, and conductivity
  • Ideal for healthcare hygiene checks
  • Compact and durable instruments with high IP protection
  • Traceable factory calibration certificate at –20 °C, 0 °C and +60 °C
The SL1011 set by ebro is a complete solution for monitoring bedpan washers and washers and disinfectors crucial in healthcare environments to ensure compliance with hygiene standards. This set includes the EBI 12-T101 temperature data logger, PHX800 pH tester, and TDS3 conductivity tester, covering critical parameters: temperature, pH, and conductivity. The EBI 12-T101 accurately logs temperature data, while the PHX800 and TDS3 provide reliable pH and conductivity measurements, ensuring comprehensive quality checks. The instruments are compact, robust, and user-friendly, designed to handle frequent use and withstand challenging conditions.
Dimensions: Ø 48×24 mm H