1-(4-Nitrophenyl)glycerol

Leverancier: Roth Carl
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synoniemen: PNPG, 1-(4-Nitrophenyl)-1,2,3-propanetriol

9240.1 9240.2
ROTH9240.1EA 0 EUR
ROTH9240.1 ROTH9240.2
1-(4-Nitrophenyl)glycerol
1-(4-Nitrophenyl)glycerol
Formule: C₉H₁₁NO₅
Smeltpunt: 94…97 °C
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00017037
CAS nummer: 2207-68-3
EINECS: 218-624-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR