1-(2-Thenoyl)-3,3,3-trifluoroacetone

Leverancier: Thermo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

SB00599.1 SB00599.10 SB00599.50
MAYBSB00599.1EA 0 EUR
MAYBSB00599.1 MAYBSB00599.10 MAYBSB00599.50
1-(2-Thenoyl)-3,3,3-trifluoroacetone
1-(2-Thenoyl)-3,3,3-trifluoroacetone
Formule: C₈H₅F₃O₂S
Kookpunt: 227 °C (1013 hPa)
Smeltpunt: 40…47 °C
Vlampunt: 111 °C
MDL Nummer: MFCD00005445
CAS nummer: 326-91-0
EINECS: 206-316-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR