4-Hydroxyphthalic acid 98%
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
412130050 412130250
ACRO412130050EA 93.4 EUR
ACRO412130050 ACRO412130250
4-Hydroxyphthalic acid 98%
4-Hydroxyphthalic acid
Formule: C₈H₆O₅
Kookpunt: 386 °C (1013 hPa)
Smeltpunt: 191…194 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00013984
CAS nummer: 610-35-5
EINECS: 210-221-6
Order Now
Specification Test Results
Appearance White to light brown Powder
Infrared spectrum Conforms
Titration with NaOH ≥97.5 %