Tanks, cilindrisch, voor zwaar gebruik, met deksel, Nalgene®
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
11100-0005 11100-0007 11100-0010 11100-0015 11100-0030 11100-0055 11100-0080 11100-0100 11100-0150 11100-0200
NALG11100-0005EA 187 EUR
NALG11100-0005 NALG11100-0007 NALG11100-0010 NALG11100-0015 NALG11100-0030 NALG11100-0055 NALG11100-0080 NALG11100-0100 NALG11100-0150 NALG11100-0200
Tanks, cilindrisch, voor zwaar gebruik, met deksel, Nalgene®
Reservoirs Cilindrische tanks
Heavy-duty cylindrical LLDPE tanks are offered in sizes ranging from 19 to 757 liters and include covers to signficantly reduce evaporation and contamination.
  • Good temperature tolerance
  • With factory-installed spigots
  • Not autoclavable
Informatie over toebehoren: Flexible tank liners: Coex polyethylene film tank liners offer cost-effective, single-use biopharmaceutical and diagnostic reagent fluid processing with rigid Nalgene® cylindrical plastic tanks
Designed to fit Nalgene® cylindrical tanks from 5 gal (19 liters) to 200 gal (757 liters)
Gamma-irradiated, food-grade film is noncytotoxic and certified nonpyrogenic
Coex polyethylene film is free of animal-derived components
Gamma irradiated 20 - 40 kGy