Real-time PCR-platen, 96-wells, Eppendorf twin.tec® 96
Leverancier: EPPENDORF
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
0030132.505 0030132.513 0030132.530 0030132.548 0030132.700 0030132.718 0030129.644 0030129.636
732-2366EA 194 EUR
732-2366 732-2367 732-2369 732-2370 732-2372 732-2373 EPPE0030129.644 EPPE0030129.636
Real-time PCR-platen, 96-wells, Eppendorf twin.tec® 96
Microplaten PCR-platen
Eppendorf twin.tec® real-time PCR plates, designed for low-volume qPCR, utilise white wells to enhance fluorescent light reflection, improving sensitivity and reproducibility in assays for reliable results.
  • White wells for better reflection
  • High mechanical stability
  • Effective sealing by raised well rims
  • Skirted and semi-skirted frames
  • Optimal heat transfer due to reduced wall thickness
  • Autoclavable (121 °C, 20 min)
  • Available in lot-certified PCR clean and Eppendorf Forensic DNA Grade for tested purity
The limiting factor in low-volume real-time PCR (qPCR) is often the remaining fluorescence intensity. Eppendorf twin.tec real-time PCR plates are specially manufactured for qPCR, as they offer all the advantages of regular twin.tec plates, but with the added advantage of white wells. These reflect fluorescence much better than clear or frosted wells, so lower levels of fluorescence are still detectable with the same instrument - just by using the right consumables. The qPCR plates white wells also significantly reduce interfering background fluorescence and lead to increased consistency between replicates, and giving highly reproducible results.
The wells are made from polypropylene - which guarantees fast heat transfer for fast assays - and the frame is made of rigid polycarbonate, to ensure easy and reliable handling - both in manual and automated assays.
Autoclavable: 121 °C, 20 min.
Centrifugation stability: 6000×g