Test Papers, Potassium Iodide Starch Papers
Leverancier: MACHEREY-NAGEL
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
90754 90755
MANA90754EA 17.3 EUR
MANA90754 MANA90755
Test Papers, Potassium Iodide Starch Papers
Testpapieren Indicatorpapier
The 816 N version comes in a classic test paper reel. You can choose the required test paper length simply by tearing the paper. Thus, MN 816 N is especially suited to be dipped into solutions.
  • Colour reaction: White to blue-violet
Order Now
Specificaties
Environmentally Preferable