Boron trifluoride - methanol complex 10% methanol, Supelco®
Leverancier: Merck
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
MQ100
15716-100ML 15716-10ML 15716-1L 15716-250ML 15716-1ML
15716-100MLEA 64.9 EUR
15716-100ML 15716-10ML 15716-1L 15716-250ML 15716-1ML
Boron trifluoride - methanol complex 10% methanol, Supelco®
Boron trifluoride - methanol complex
Formule: CH₄BF₃O
MW: 99.85 g/mol
Dichtheid: 1.222
MDL Nummer: MFCD00071635
CAS nummer: 373-57-9
EINECS: 206-766-4
UN: 3286
ADR: 3 II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=612f76cb6db91dcf7502c059b61da9608e7730d7&vid=9a47800cc30d1a2cd0d48bc41fdacc864dfce83f&prid=web&clid=SAPDAM