Tanks, rechthoekig, met deksel, Nalgene®
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
14100-0005 14100-0010 14100-0015 14100-0020 14100-0040 14100-0045 14100-0065
216-1912EA 377 EUR
216-1912 216-1913 216-1914 216-1915 NALG14100-0040 NALG14100-0045 NALG14100-0065
Tanks, rechthoekig, met deksel, Nalgene®
Reservoirs Rechthoekige tanks
These heavy-duty rectangular LLDPE tanks with cover can withstand most acid, alcohols and bases. Tanks feature stepped flange at top provides reinforcement and grip for lifting.
  • Impact- and abrasion-resistant
  • Molded as one piece, no seams
  • Nominal wall thickness is 3,97 mm
  • Stepped flange at top provides reinforcement and grip for lifting
  • Not autoclavable