Size exclusion chromatography columns for proteins and antibodies, Shodex, PROTEIN KW and LW series
Leverancier: Resonac
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
F6989000 F6989202 F6989203 F6700132 F6989201 F6989204 F6989104 F6989103 F6700131 F6700133 F6989303 F6989403 F6700134
554-1735EA 1652 EUR
554-1735 554-1741 554-1742 554-1744 554-1740 554-1743 554-1737 554-1736 554-1739 554-5779 554-5778 SHOWF6989403 SHOWF6700134
Size exclusion chromatography columns for proteins and antibodies, Shodex, PROTEIN KW and LW series
Kolommen voor chromatografie
PROTEIN KW-800 and KW400 series HPLC columns are specifically designed for protein analysis by aqueous size exclusion chromatography (GFC). PROTEIN LW-803 and LW-403 4D are designed for antibody analysis. They are packed with porous silica particles coated with a hydrophilic copolymer. They fulfill USP L20, L33, and L59 requirements.
Standard columns PROTEIN KW-800 series
- Silica-based packing material for aqueous SEC analysis.
- Suitable for the analysis of proteins and enzymes.
- There are three pore sizes available for different molecular weight ranges.
High performance semi-microMore Product Information