Reductiestuk met kegelslijping
Leverancier: SciLabware
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
DA35 DA12 DA23 DA14 DA24 DA34 DA13 DA45 DA01 DA25 DA58 DA36 DA38 DA56 DA37 DA47 DA57
201-2375EA 88.4 EUR
201-2375 201-2357 201-0111 201-8344 201-8346 201-8349 201-0255 201-0150 201-0639 201-0112 201-0644 BIBBDA36 BIBBDA38 BIBBDA56 BIBBDA37 BIBBDA47 BIBBDA57
Reductiestuk met kegelslijping
Adapters Adapters met NS-verbindingen
PYREX®, borosilicate glass, Quickfit® joints, clear.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Reduction adapter from large cone at the base to small socket on the top.