Antistatische spray, Antistatik 100
Leverancier: HM INDUSTRIESERVICE
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
100085
291-1240EA 20.2 EUR
291-1240
Antistatische spray, Antistatik 100
Reinigingsmiddelen Detergenten en oppervlaktereinigers
ANTISTATIK 100 forms an invisible protective film against electrostatic charges. The antistatic solution with cleaning alcohol removes dirt and offers long-lasting protection against static electricity. ANTISTATIK 100 cleans and protects plastic housings, textile surfaces, screens, acrylic glass covers or packaging films.