Bags, zip seal, kangaroo pocket, Minigrip®
Leverancier: Flexico
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
ITM020811 ITM020813 ITM020812
129-2013EA 348 EUR
129-2013 129-2015 300-0227
Bags, zip seal, kangaroo pocket, Minigrip®
Zakken Monsterzakken
Clear PE bags, thickness 60 µm, food contact.
  • Extra external pocket to protect accompanying documents
  • Suitable for food contact
  • Hanging hole
  • Resealable, reusable, 100% recyclable
Made of polyethylene (LDPE). Zip seal designed for easy opening and high speed packing. The Minigrip® bag provides an excellent mechanical resistance and high temperature resistance (from –40 up to +60 °C).
Certificaten: Suitable for food contact. Certificate available on request.