Buisfolie voor sterilisatie
Leverancier: SÜDPACK MEDICA
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
7600547 7600513 7600515 7600518 7600519 7600523 7600526 7600532 7600538 7600546 7600567 7600508
129-1086EA 465 EUR
129-1086 129-1422 129-1423 129-1434 129-1425 129-1426 129-1435 129-1411 129-1412 129-1430 129-1437 129-0485
Buisfolie voor sterilisatie
Zakken Sterilisatiezakken
Tubular bag film is available as a flat or gussetted bag in various sizes. Helpful tips for use are printed on the film. Suitable for autoclaving and ethylene oxide sterilisation.
* Afmeting van de balg