Zakken, biologisch gevaarlijk, SP Bel-Art
Leverancier: Bel-Art Products, a Part of SP
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
F131641419 F131641923 F131642535 F131643138 F131643848 F131651419 F131651923 F131652535 F131653138 F131653748
129-1000EA 335 EUR
129-1000 129-1001 129-1002 129-1003 129-1004 129-1005 129-1006 129-1007 129-1061 129-1008
Zakken, biologisch gevaarlijk, SP Bel-Art
Zakken Afvalzakken
Bags, in PP, 38 or 50 µm thick, orange, with 'Biohazard' symbol.
  • Printed sterilisation patch that darkens when subjected to steam sterilisation
  • Black biohazard symbol
Type A: PP bags autoclavable to 135 °C, thickness: 38 µm.
Type B: Extra heavy duty bags blended, autoclavable to +121 °C, thickness: 50 µm.