Afvalzakken, Biohazard
Leverancier: Alpha Packaging
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
121-075 121-076 121-077 121-078
129-0354EA 119 EUR
129-0354 129-0355 129-0356 129-0357
Afvalzakken, Biohazard
Zakken Afvalzakken
Bags with printed black symbol for biological risk material on yellow background and with line for max. filling.
PE, thickness 500, transparent, with label 'Biohazard'.
  • Each bag is box shaped
  • Double string neck pull for easy closure