Afvalzak, Biohazard, Hi-Temp
Leverancier: Alpha Packaging
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
121-100 121102
129-0413EA 0 EUR
129-0413 129-0414
Afvalzak, Biohazard, Hi-Temp
Zakken Afvalzakken
Bags with black warning symbol on yellow background. PP, 62,5 µm (250 gauge), transparent, with 'Biohazard' label
  • Safety use instructions printed on each bag
  • Suitable for temperatures up to 135 °C
Verpakking: Supplied in benchtop dispenser boxes.
Order Now
Specificaties
Environmentally Preferable