Afvalzakken, Biohazard, zelfklevend
Leverancier: Alpha Packaging
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
121103 121104 121105
129-1931EA 172 EUR
129-1931 129-0398 129-0399
Afvalzakken, Biohazard, zelfklevend
Zakken Afvalzakken
Tough, yellow biohazard bags, non autoclavable. Supplied in benchtop dispenser boxes. PE, 62,5 µm (250 gauge).
  • Peel-off permanent adhesive seal
  • Remove backing and seal for safer transportation/disposal
  • Biohazard warning in English, German and French, black printing on yellow background