Zak voor stralingsafval
Leverancier: Alpha Packaging
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
121-067 121-068 121-069BOX 121-071 121-033 121-034 121-035 121-070 121-037
129-0346EA 66.8 EUR
129-0346 129-0347 APHA121-069BOX 129-0350 129-0343 129-0344 129-0348 129-0349 129-0345
Zak voor stralingsafval
Zakken Afvalzakken
Bags, printed on one side; yellow centre with black trefoil and the wording 'Caution Radioactive Material'. HDPE, 125 µm, transparent
  • Warning given in English, French and German
  • Supplied in benchtop dispenser boxes or loose packs