Autoclaveerbare zakken
Leverancier: Greiner Bio-One
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
643401_500 644201_500 646401_300 649401_200 644401_500
129-9721EA 339 EUR
129-9721 GREI644201_500 129-9723 129-9724 129-9722
Autoclaveerbare zakken
Zakken Afvalzakken
These bags are designed specifically for the disposal of contaminated laboratory waste, that first requires steam sterilisation.
  • Bags for autoclaving and oven - resistant up to 160 ° C in an oven and 134 ° C in an autoclave
  • Suitable for hot air sterilisers
Order Now
Specificaties
Environmentally Preferable