Sterilisatiemanden voor proefbuizen
Leverancier: Bel-Art Products, a Part of SP
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
F187370010 F187380010 F187390010 F187370000
489-2210EA 91.3 EUR
489-2210 489-2211 489-2212 489-2230
Sterilisatiemanden voor proefbuizen
Manden Reageerbuismanden
PP.
  • Very good chemical resistance
  • Very good temperature resistance
  • Sterilisable and autoclavable
These test tube baskets are designed for rinsing, transporting and storing laboratory glassware.
Colour: White