Reageerbuismanden
Leverancier: SciLabware
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
BWK006 BWK009
215-0549EA 79 EUR
215-0549 215-0550
Reageerbuismanden
Manden Reageerbuismanden
PP.
  • Very good chemical resistance
  • Autoclavable at 135 °C
  • Highly break-resistant
  • Stackable space saving design
These tests tubes baskets are suitable for rinsing, sterilising, transporting and storing glassware.
Colour: White