Wire baskets
Leverancier: HUPFER METALLWERKE
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
7500900 7500901 7500902 7500903 7500904 7500905 7500906 7500907 7500929
215-7411EA 91.4 EUR
215-7411 215-7412 215-7414 215-7416 215-7418 215-7420 215-7422 215-7424 215-3300
Wire baskets
Manden Draadmanden
Stainless steel.
  • Spot welded mesh 8×8×1 mm
  • Electrolytically polished
  • Reinforced edges
These baskets can be used for transport, storage, cleaning and sterilisation of laboratory utensils.
Order Now
Specificaties
Environmentally Preferable