Bekers, driehoekig, Tricorn™
Leverancier: Simport Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
B700-100 B700-1L B700-250 B700-400 B700-800 B700-50
212-9302EA 61.9 EUR
212-9302 212-9306 212-9303 212-9304 212-9305 212-9301
Bekers, driehoekig, Tricorn™
Bekers
These Tricorn™ beakers are made of polypropylene, provide three dripless pouring spouts that offers more security.
  • Autoclavable
  • Tough and unbreakable
Suitable for use with commonly used acids, alkalis and solvents.
Verpakking: All sizes are available 100 per case.
Opgelet: *De product(en) op deze pagina vallen buiten het toepassingsgebied van de EU-richtlijn 2019/904