Bekers, lage vorm, Azlon®
Leverancier: SciLabware
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
BPM0050P BPM0100P BPM0500P BPM1000P BPM2000P BPM0250P
213-0107EA 25.5 EUR
213-0107 213-0108 213-0110 213-0112 213-0113 213-0109
Bekers, lage vorm, Azlon®
Bekers
Zeer doorschijnend PP.
  • Tapered stacking style - space saving and easy to handle
  • Autoclavable, durable with good chemical resistance
  • Precise non-drip pouring spouts, with easy-to-read moulded graduations
Certificaten: Comply with ISO 7056.