Klokvormige maatbekers
Leverancier: GLASWARENFABRIK KARL HECHT
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
42260012 42260042 42260018 42260024 42260048 42260030 42260006 42260036
612-2756EA 17.6 EUR
612-2756 612-2757 612-2758 612-2759 612-2760 612-2761 612-2762 612-2763
Klokvormige maatbekers
Cilinders Maatcilinders
AR glass, clear.
  • Very good chemical resistance
  • Good thermal resistance
  • Fill volume is easy to read
With a broad base for stability. Fused blue graduations, very easy to read. Class B tolerance, twice the official tolerance. No handle.