Maatbekers met handvat
Leverancier: NICKEL ELECTRO
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
JUGS0500H JUGS1000H JUGS2000H
NICKJUGS0500HEA 61.7 EUR
NICKJUGS0500H 213-0267 NICKJUGS2000H
Maatbekers met handvat
Bekers
Roestvrij staal, roestvrij.
  • Very good chemical resistance
  • Very good thermal resistance
  • Smooth surfaces for fast, easy cleaning
Conical, without rim, with broad base and textured graduations.