Filtreerbekers, DURAN®
Leverancier: DWK Life Sciences
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
211312409 211312906 211313602 211314401 211315406 211316308 211316805 211317304 211318609 211318806 211319108
511-0315EA 29.3 EUR
511-0315 511-0316 511-0340 511-0317 511-0318 511-0319 511-0320 511-0321 511-0322 511-0323 511-0324
Filtreerbekers, DURAN®
Bekers
DURAN®, borosilicaatglas 3.3, helder.
  • Increased wall thickness for better mechanical properties
  • Limited use as cooking glass due to reduced thermal shock resistance
  • Available graduated or non graduated
Dikwandig, met tuit, gemakkelijk afleesbare schaal en groot labelveld.
Above 10000 ml without graduation