Decanteerflessen met zijtubus
Leverancier: SciLabware
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
1531/08D 1531/10D 1531/12D
215-0349EA 527 EUR
215-0349 BIBB1531/10D 215-0350
Decanteerflessen met zijtubus
Flessen Doseerflessen
PYREX®, borosilicate glass, without stoppers.
  • Excellent chemical resistance
  • Very good temperature resistance
Clear with white labelling area and unground neck to accept rubber stoppers. Glass tubular side arm for attachment of 12 mm bore flexible tubing.
Order Now
Specificaties
Environmentally Preferable