Aspirator bottles, narrow mouth, HDPE, Azlon®
Leverancier: SciLabware
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
BGH034 BGH036 BGH024 BGH038 BGH026 BGH028 BGH040 BGH022
216-2780EA 100 EUR
216-2780 216-2798 216-2795 216-2799 216-2796 216-2797 216-2800 216-2794
Aspirator bottles, narrow mouth, HDPE, Azlon®
Flessen Doseerflessen
Made from PE-HD, with narrow neck and screw cap.
  • Perfectly adapted for storing in bulk, transporting liquids, and dispensing common solvents or distilled water on laboratory countertops thanks to their watertight taps
  • Fitted with carrying handles (except 5 litre bottles)
  • Good chemical resistance
Items 216-2794 to 216-2797 without a draw-off tap.
Items 216-2780 to 216-2800 with a draw-off tap.