Carboys, brede hals, PP, met schroefdop, Nalgene®
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
2250-0050 2250-0130 2250-0020
216-8762EA 176 EUR
216-8762 216-8763 216-8761
Carboys, brede hals, PP, met schroefdop, Nalgene®
Flessen Voorraadflessen
Doorschijnend PP, met witte PP-schroefdop, TPE-dichting.
  • Zeer goede chemische bestendigheid
  • Hittebestendig tot 135 °C, autoclaveerbaar
  • Extra brede opening, brede grepen op de schouders, lekvrij
Ideaal voor grote hoeveelheden voedingsbodems, gedistilleerd water of andere oplossingen.
These autoclavable carboys are used for large batches of culture media, vaccine product production, bulk media and chemical storage. These carboys feature a large mouth for easy filling/dispensing, and are made of natural polypropylene with white 83B polypropylene closure and TPE (thermoplastic elastomer) gasket.
Customisation options available for fluid transfer applications. Fit perfectly with a variety of optional 83B closures with standard tubing sets (such as Nalgene™ Top Works™, Nalgene™ Filling/Venting) and custom tubing set designs.
Convenient shoulder handles allow easy carrying and pouring, even with gloved hands. Graduated to contain in 1 gal. and 5 L increments for ease of use. These carboys are autoclavable and graduated.