Voorraads- en uitklaarflessen, Azlon®
Leverancier: SciLabware
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
BWX072 BGH068 BGH070
215-3170EA 974 EUR
215-3170 215-3168 215-3169
Voorraads- en uitklaarflessen, Azlon®
Flessen Doseerflessen
Doorschijnend HDPE.
  • Good chemical resistance, good temperature resistance from -100 up to +80 °C (for short periods up to 120 °C)
  • Leakproof screw cap
With white screw cap and tightly fitting drain cock. Extra wide opening for easy filling. Ideal for storage and transport as well as for dispensing liquids. Also recommended for use as aspirator bottles. With two carrying handles.