Carboys, brede hals, LDPE, met schroefdop, Nalgene®
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
2210-0020 2210-0040 2210-0050 2210-0065 2210-0130
216-4393EA 70.6 EUR
216-4393 216-8702 216-4394 216-4395 216-4396
Carboys, brede hals, LDPE, met schroefdop, Nalgene®
Flessen Voorraadflessen
LDPE, translucent with white PP screw cap.
  • Good chemical resistance
  • Heat resistant up to +80 °C
  • Extra wide opening, broad shoulder handles make carrying and pouring easier, even when wearing gloves, leakproof closure
Ideal for storing and transporting reagents that react readily with HDPE polymerisation catalysts. Graduated.