Bronflessen, brede hals, PP, met schroefdop en buisconnector, Nalgene®
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
2301-0020 2301-0050
215-0118EA 91.2 EUR
215-0118 215-0119
Bronflessen, brede hals, PP, met schroefdop en buisconnector, Nalgene®
Flessen Voorraadflessen
PP, translucent with white PP screw closure and tube connector for tubes with 13 mm internal diameter.
  • Very good chemical resistance
  • Thermal resistance from -10 to +135 °C, autoclavable
  • Leakproof closure
The tube connector is moulded in one piece with the carboy for improved stability and to provide a better seal. The outlet is set as low as possible so that the carboy can be emptied completely. Convenient, wide shoulder handles for easy carrying. Graduated.
Bestelinformatie: Supplied without tubing.