Carboys, smalle hals, met schroefdop, Azlon®
Leverancier: SciLabware
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
BNP05B BNP10B BNP20B
216-3651EA 51.6 EUR
216-3651 216-3652 216-3653
Carboys, smalle hals, met schroefdop, Azlon®
Flessen Voorraadflessen
PP, translucent with white PP screw cap.
  • Very good chemical resistance
  • Heat resistant up to 135 °C, autoclavable
  • Extra wide opening, with handle on the shoulders
Ideal for large quantities of culture media, distilled water or other solutions. With graduations.