Voorraads- en uitklaarflessen
Leverancier: hünersdorff GmbH
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
823300 823400 823500
216-5841EA 41.4 EUR
216-5841 216-5842 216-5843
Voorraads- en uitklaarflessen
Flessen Voorraadflessen
Carboys, HDPE, white, deal for storing and transporting as well as dispensing liquids. Also recommended for use as aspirator bottles. With metal carrying handles.
  • Good chemical resistance, good temperature resistance from -100 to 80 °C (up to 120 °C for brief periods)
  • With wide threads and threaded stopper for connecting a valve cock (please order 216-5291 or 216-5292 separately)
  • Leakproof screw cap